Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Magnetic-Ring Stand (96 well)

Catalog number: AM10050
Have Questions?
Invitrogen™

Magnetic-Ring Stand (96 well)

Catalog number: AM10050
Have Questions?
Catalog Number
AM10050
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM100501 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ambion™ 96-well Magnetic-Ring Stand is designed for paramagnetic bead precipitation from standard 96-well U-bottom microplates. The stand has a metal base in a standard SBS 96-well microtiter plate footprint. The use of 96 extremely strong permanent magnetic rings enables fast and easy magnetic separation. With this magnetic stand, paramagnetic beads in a U-bottom plate will pellet in a donut shape ∼ 6 mm in diameter, which enables easy removal of supernatant by a robot or with a multichannel pipettor. It sits flat on a lab bench, as well as in a microtiter plate adapter on robotic workstations.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Magnetic Stand
Shipping Condition
Room Temperature
Product Line
Ambion™
Quantity
1 Each
Type
Magnetic Stand

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text