Spin Columns and Tubes
Spin Columns and Tubes
Invitrogen™
Spin Columns and Tubes
The Ambion® Spin Columns are for the rapid elution of DNA from agarose gel slices. No electroelution is needed; theRead more
Have Questions?
Catalog NumberQuantity
AM10065100 units
Catalog number AM10065
Price (USD)
297.65
Online Exclusive
311.00
Save 13.35 (4%)
Each
-
Add to cart
Quantity:
100 units
Delivery information
Standard Products - 1-2 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 2-3 business days for 2nd-tier cities, and 4 business days for 3rd-tier cities and remote areas.
Air Freight Restricted Products - 2-4 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 3-5 business days for 2nd-tier cities, and 6-10 business days for 3rd-tier cities and remote areas.
Supply Center: Delivered from the Supply Center closest to you.
Price (USD)
297.65
Online Exclusive
311.00
Save 13.35 (4%)
Each
Add to cart
Ask our AI about this Product
The Ambion® Spin Columns are for the rapid elution of DNA from agarose gel slices. No electroelution is needed; the columns only require a microcentrifuge. The columns are certified DNase- and RNase-free. 100 units are provided. Simply excise the band of interest from the gel, place it in the spin column, and centrifuge for 5 min! The DNA is forced through the column into the tube below while the agarose is left behind. The DNA is then ready for subsequent manipulations such as restriction enzyme digestion or ligation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)DNA Extraction from Agarose Gels
Product TypeSpin Columns and Tubes
Quantity100 units
Shipping ConditionRoom Temperature
DisposableSingle-use
FilteredFiltered
For Use With (Equipment)Microcentrifuge
FormatTube, Spin Column
Product LineInvitrogen
Tip StyleUniversal Fit
Unit SizeEach
Contents & Storage
Store tubes at room temperature.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
浏览此商品的顾客同时也查看了
Documents & Downloads
Certificates
Frequently asked questions (FAQs)
Yes, we recommend Spin Columns and Tubes (Cat. No. AM10065) (https://www.thermofisher.com/order/catalog/product/AM10065).
We recommend using Spin Columns and Tubes (Cat. No. AM10065): https://www.thermofisher.com/order/catalog/product/AM10065
Share catalog number, name or link
1x1 image pixel for data collection