The Retic Lysate IVT™ Kit efficiently translates in vitro-synthesized transcripts, poly(A) RNA, and total RNA, including difficult-to-translate RNAs. The optimizedRead more
Have Questions?
Change view
Catalog Number
Product Type
Quantity
AM1200M
Retic Lysate
60 Reactions
AM1200
Retic Lysate
60 Reactions
Catalog number AM1200M
Price (USD)/ 60 reactions
835.65
Online exclusive
860.00
Save 24.35 (3%)
-
Add to cart
Product Type:
Retic Lysate
Quantity:
60 Reactions
Request bulk or custom format
Price (USD)/ 60 reactions
835.65
Online exclusive
860.00
Save 24.35 (3%)
Add to cart
Retic Lysate IVT™ Kit
Catalog numberAM1200M
Price (USD)/ 60 reactions
835.65
Online exclusive
860.00
Save 24.35 (3%)
-
Add to cart
The Retic Lysate IVT™ Kit efficiently translates in vitro-synthesized transcripts, poly(A) RNA, and total RNA, including difficult-to-translate RNAs. The optimized lysate results in high protein yield, biological activity, and consistent performance. Advantages of the Retic Lysate IVT™ Kit:
• Efficiently translates a wide variety of high molecular weight proteins and both capped and uncapped mRNAs • Accommodates large volume of template mRNA
How it works The lysate has been optimized for translation by the addition of an ATP-regenerating system, hemin, and calf liver tRNA. Included in the kit is an optimized Translation Mix (-Met or -Leu) for the translation of both capped and uncapped messages along with High and Low Salt buffers. These buffers can be used to maximize protein yield and activity from difficult-to-translate sequences. A report in Nucleic Acids Research (1) found that some commercial reticulocyte lysates had poor fidelity for initiation at the appropriate AUG initiator codon. The Retic Lysate IVT™ Kit was tested using Kozak's constructs and it demonstrated high fidelity for initiating at the proper AUG.
Reference 1. Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res 18:2828.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
Expressed Protein TypeWater Soluble Protein
Expression MechanismCell-Free Expression
Expression SystemCell-Free
For Use With (Application)In Vitro Transcription
No. of Reactions60 Reactions
Product LineAmbion™
Product TypeRetic Lysate
Quantity60 Reactions
Unit Size60 reactions
Contents & Storage
Retic Lysate should be stored at –80°C. 20X Translation Mix minus-Met, 20X Translation Mix minus-Leu, High Salt Mix minus-Met, Low Salt Mix minus-Met, and Capped Xef-1 RNA should be stored below –70°C. Nuclease-Free water can be stored at any temperature.