Invitrogen™

MEGAscript™ T7 Transcription Kit

Catalog number: AM1333
Have Questions?
Invitrogen™

MEGAscript™ T7 Transcription Kit

Catalog number: AM1333
Have Questions?
Catalog Number
AM1333
Unit Size
25 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
40 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM133325 reactions
-
AMB13345200 reactions
-
AM133440 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The MEGAscript™ T7 Kit is an ultra-high yield in vitro transcription kit. The high yields are achieved by modifying typical transcription reaction conditions so that very high nucleotide concentrations can be effectively used. The T7 Enzyme Mix and the 10X Reaction Buffer are specifically calibrated for each lot and RNA polymerase.

Using the MEGAscript™ T7 Kit
The MEGAscript™ T7 Kit contains in vitro transcription reaction components and a control template. The kit will yield a total of 3–5 mg of RNA (approximately 100 μg of RNA or more per reaction) from the control template supplied with the kit. This corresponds to 400–650 moles of RNA for each mole of template. Smaller templates typically yield a lower mass and a higher molar yield of product. Novel transcription reaction conditions and a patented, high-yield technology allow the synthesis of 10–50 times the amount of RNA produced by conventional transcription reactions.

Applications
The MEGAscript™ T7 Kit is intended for the synthesis of large amounts of unlabeled or low specific activity RNA for a variety of uses, including in vitro translation, antisense/microinjection studies, and isolation of RNA binding proteins. In large-scale transcription reactions, the concentration of all 4 nucleotides is high, well above the Km for the enzyme. The MEGAscript™ T7 Kit typically yields over 10 times more RNA than conventional in vitro transcription reactions. The kit is not recommended for synthesis of high specific activity probes.

Accessory products
MessageAmp™ aRNA amplification kits are powered by MEGAscript™ technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, we recommend using the MEGAclear™ Kit (Cat. No. AM1908) and the MEGAclear™-96 Kit (Cat. No. AM1909).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product Type
Synthetic mRNA, mRNA
Format
Kit
Sample Type
mRNA, Synthetic mRNA
For Use With (Application)
In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions
25 Reactions
Product Line
Ambion™, MEGAscript™
Product Type
T7 In Vitro Transcription kit
Quantity
25 Reactions
Promoter
T7
Shipping Condition
Dry Ice
Yield
100 μg
Starting Material
PCR Products, Linearized DNA plasmid

Contents & Storage

T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text