Invitrogen™

Poly(A) Tailing Kit

Catalog number: AM1350
Have Questions?
Invitrogen™

Poly(A) Tailing Kit

Catalog number: AM1350
Have Questions?
Catalog Number
AM1350
Unit Size
25 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM135025 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
For the polyadenylation of in vitro transcribed RNA to enhance translation initiation efficiency. Sufficient reagents are included for 25 reactions.

• Adds a poly(A) tail of at least 150 nucleotides in length to the 3' termini of RNA
• Enhances translational efficiency in vivo
• Reaction can be adjusted to control for tail length
• Optimized for use with RNA transcripts synthesized using the mMESSAGE mMACHINE™ Kit

The Poly(A) Tailing Kit uses E. coli Poly(A) Polymerase I (E-PAP) to polyadenylate the 3'-termini of in vitro transcribed RNA. Polyadenylation plays an important role in the stabilization of RNA in eukaryotes and enhances the efficiency of translation initiation. In the Poly(A) Tailing Kit, the E-PAP reaction has been optimized so that mRNAs are efficiently tailed with at least 150 adenines. The additional adenine residues confer stability to the mRNA and may increase translational efficiency of in vitro synthesized capped RNA in microinjection and transfection experiments (see Figure).

Accessory Products:
The Poly(A) Tailing Kit is optimized for use with Ambion's mMESSAGE mMACHINE™ High Yield Capped RNA Transcription Kit. mMESSAGE mMACHINE™ Kits are available with T7 (SKU #AM1334), T3 (SKU #AM1338), or SP6 (SKU #AM1330) polymerases.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Kit Type
Tailing Kit
Product Line
Ambion™
Shipping Condition
Dry Ice
For Use With (Application)
miRNA analysis
Final Product Type
In Vitro Transcribed RNA (Polyadenylated)
No. of Reactions
25 Reactions
Starting Material
RNA (In Vitro Transcribed)
Unit Size
25 reactions

Contents & Storage

E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, and Control DNA Template should be stored at -20°C.

Nuclease-free Water may be stored at any temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text