Pre-miR™ miRNA Starter Kit
Product Image
Pre-miR™ miRNA Starter Kit
Have Questions?
Catalog number AM1540
Price (USD)/ 1 kit
ERR#1200 - This item cannot be ordered online.
Ambion® Pre-miR miRNA Starter Kit is a complete solution for controlling and transfection optimization in miRNA gain-of-function experiments mediated by synthetic miRNA mimics. It contains a positive control, a TaqMan® Gene Expression Assay to measure the positive control, a negative control, and a transfection agent.
• Get started analyzing miRNA function via gain-of-function experiments
• Includes positive and negative control Pre-miR™ miRNA Precursors, a matched real-time PCR assay, and transfection agent
• Accelerates transfection optimization of miRNA mimics into human cells
Upon delivery into cells, the positive control (miR-1 Pre-miR miRNA Precursor) effectively down regulates the expression of twinfilin-1, also known as PTK9, at the mRNA level. This can be detected by real-time PCR using a TaqMan® Gene Expression Assay for PTK9 (Assay ID: Hs00702289_s1), which is included in the kit. (Note: the control miRNA mimic can also be used in mouse cells, but requires a specific TaqMan® Gene Expression Assay for mouse PTK9 that is available separately; Assay ID Mm01598980_g1.)
The included negative control does not target any known mRNA within the human or mouse transcriptome. Also included is siPORT™ NeoFX™ Transfection Agent, a lipid-based transfection agent optimized for use with siRNAs and miRNA mimics.
Specifications
Unit Size1 kit
Contents & Storage
Pre-miR™ hsa-miR-1 miRNA Precursor, Pre-miR™ Negative Control #1, and Hs00702289_s1 20X PTK9 (PCR Primer⁄Probe Set) should be stored at –20°C.
siPORT™ NeoFXTransfection Agent should be stored at 4°C, and Nuclease-free Water may be stored at any temperature.