The Ambion™ QuantumRNA™ 18S Internal Standard with Competimer™ technology uses specially modified Competimer™s of the same sequence as the normalRead more
Have Questions?
Catalog number AM1717
Price (USD)/ 100 reactions
288.00
-
Add to cart
Price (USD)/ 100 reactions
288.00
Add to cart
QuantumRNA™ Classic II 18S Internal Standard
Catalog numberAM1717
Price (USD)/ 100 reactions
288.00
-
Add to cart
The Ambion™ QuantumRNA™ 18S Internal Standard with Competimer™ technology uses specially modified Competimer™s of the same sequence as the normal 18S primers that cannot be extended. By adjusting the ratio of 18S Competimer™s to normal 18S mRNA primers, the signal for 18S mRNA can be attenuated to the level of very rare messages. They are used for quantitative end-point RT-PCR and include sufficient reagents for 100 reactions. Because of its invariant expression across tissues and treatments, 18S ribosomal RNA is an ideal internal control for quantitative RNA analysis. The Classic II 18S Internal Standards produce a 324 bp PCR product.
Accessory Products: The QuantumRNA™ Classic 18S Internal Standard (SKU# AM1716) produce a 489 bp PCR product, and the QuantumRNA™ Universal 18S Internal Standard (SKU# AM1718) function across the broadest range of eukaryotes, including animals, plants, and many protozoa and produce a 315 bp PCR product.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Control TemplateTotal RNA from Mouse Liver
FormatTube(s)
Gene Symbol18S
Label or DyeUnlabeled
Product LineAmbion™, QuantumRNA™
Shipping ConditionDry Ice
SpeciesMouse
Target18S rRNA
No. of Reactions100 Reactions
Unit Size100 reactions
Contents & Storage
18S Primer Pair and 18S Competimer™s are stored at –20°C. Control Template RNA should be stored below –70°C. Nuclease-free water may be stored at any temperature.