Cells-to-cDNA™ II Kit
Cells-to-cDNA™ II Kit
Invitrogen™

Cells-to-cDNA™ II Kit

The Ambion™ Cells-to-cDNA™ II Kit (patent pending) produces cDNA from cultured mammalian cells in less than 2 hr. No RNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AM172240 reactions
AM1723100 reactions
Catalog number AM1722
Price (USD)
684.65
Online Exclusive
734.00
Save 49.35 (7%)
Each
Add to cart
No. of Reactions:
40 reactions
Price (USD)
684.65
Online Exclusive
734.00
Save 49.35 (7%)
Each
Add to cart

The Ambion™ Cells-to-cDNA™ II Kit (patent pending) produces cDNA from cultured mammalian cells in less than 2 hr. No RNA isolation is required. This kit contains sufficient reagents for 100 reactions and is ideal for those who want to perform reverse transcription reactions on small numbers of cells, numerous cell samples, or for scientists who are unfamiliar with RNA isolation.

• No RNA purification required
• From cells in culture to cDNA in less than 2 hr
• Detect rare messages in as few as 1 cell
• Ideal for labs not equipped for RNA isolation

Ideal for RT-PCR Applications
The Cells-to-cDNA™ II Kit (patent pending) integrates RNase inactivation and DNase I treatment into an RT-PCR compatible cell lysis buffer, eliminating RNA isolation altogether. Most cell lysis buffers contain strong denaturants that if carried over into enzymatic reactions would inhibit or inactivate most enzymes. If a lysis buffer without strong denaturants is used, endogenous RNase can quickly degrade cellular RNA. The Cells-to-cDNA™ II Kit contains a novel Cell Lysis Buffer that inactivates endogenous RNases without compromising downstream enzymatic reactions. After inactivation of RNases, the cell lysate can be directly added to a cDNA synthesis reaction. Cells-to-cDNA™ II is compatible with both one-step and two-step RT-PCR protocols.

Quantitative—Linear Results and No Detectable DNA Contamination
To illustrate the quantitative, linear response of Cells-to-cDNA™ II to variations in input cell number, a real-time quantitative RT-PCR experiment was performed using the ABI 7700 Sequence Detection System. Plotting cycle threshold (Ct) versus cell concentration for GAPDH yielded a standard curve with a correlation of 0.99. Thus, Cells-to-cDNA™ II yields linear results for 1 to 10,000 cells/ μL in a real-time RT-PCR two-step assay. It should also be noted that the minus-template PCR control for the GAPDH experiment was negative, indicating complete removal of genomic DNA.

Simple Procedure
The kit comes with everything you need to go from cultured cells to PCR-ready cDNA in less than 2 hr; no RNA isolation is required. Cells are first washed with PBS and then resuspended in the Cell Lysis Buffer. A brief heating simultaneously lyses the cells and inactivates RNases. An optional DNase digestion is then performed to degrade genomic DNA, followed by a heat inactivation step. A fraction of the cell lysate is then used in a reverse transcription reaction with the included reagents.

Accessory Products:
SuperTaq™ Thermostable Taq DNA Polymerase is recommended and available separately (Cat. No. AM2050 and AM2052). For optimal amplification of fragments greater than 1 kb, use SuperTaq™ Plus Thermostable Taq DNA Polymerase (Cat. No. AM2054 and AM2056).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypecDNA
No. of Reactions40 reactions
Optimal Reaction Temperature42°C
Product LineAmbion™, Cells-to-cDNA™
Purification Time2 hr.
Quantity40 reactions
Reverse TranscriptaseM-MLV
Sample TypeMammalian Cells
Shipping ConditionDry Ice
Size (Final Product)7 kb or less
For Use With (Application)Real Time PCR (qPCR), RT-PCR
Unit SizeEach
Contents & Storage
1X PBS, Cell Lysis II Buffer, μL DNase 1, 10X RT Buffer, M-mLV Reverse Transcriptase, RNase Inhibitor, dNTP Mix, Random Decamers, Oligo(dT)18 Primers, Armored RNA™ Control, Armored RNA™ Primer Pair, and Endogenous Primer Pair should be stored at
-20°C. Nuclease-free water may be stored at any temperature.
Please enter sku to load default questions

Frequently asked questions (FAQs)

I'm seeing PCR products in the minus-RT control after performing my Cells-to-CT experiment. What does this mean?

If PCR products are seen in the minus-RT control reaction, but not in the no-template control, it indicates that genomic DNA remains in the sample and that genomic DNA was amplified in real-time PCR. Please follow the suggestions below:

- Ensure the DNase I is mixed thoroughly into the Lysis Solution.
- Use fewer cells per lysis reaction.
- Lyse cells using Lysis Solution that is at room temperature, and make sure that the lysis reaction occurs at room temperature.
You can also try increasing the incubation time of the lysis reaction to 8 minutes and/or using Lysis Solution that has been warmed up to 25 degrees C for cell lysis.

I'm getting PCR products in the no-template PCR control when performing a Cells-to-CT experiment. What could cause this?

PCR products in the no-template PCR control indicate that the sample is contaminated with DNA. More stringent steps need to be taken to control contamination.

I'm getting no PCR product or unexpected PCR products after performing a Cells-to-CT experiment. What could be the cause of this?

Please review the following possibilities and suggestions:

- A problem with adding or mixing the Stop Solution: ensure that the Stop Solution was added directly to the lysate, as components of the Lysis Solution may inhibit RT-PCR if not fully inactivated.
- The RNA was degraded: keep cells in PBS on ice before starting the cell lysis procedure.
- RNase in the sample was not completely inactivated: Too many cells could have been used or too much PBS left on the cells, diluting the lysis solution.
- The lysates sat too long before going to room temperature: Do not allow lysates to sit longer than 20 minutes at room temperature once the Stop Solution has been added.
- The sample does not contain the target RNA: Verify that the procedure is working by using the XenoRNA Control in the sample. Also check that your PCR primers can amplify your target under the PCR conditions you are using.

I ran out of stop solution for my Cells-to-CT experiment. Can I purchase it separately?

Yes, it is available in 1 mL aliquots (Cat. No. 4402960).

I have genomic DNA contamination in my Cells-to-CT reaction. How do I get rid of it?

1. Ensure that all medium is removed from the wells.
2. Wash with an equal volume of room temperature 1X PBS after the medium is removed.
3. Ensure that the reaction happens at room temperature (the lysis reaction may not reach room temperature if the plate is on ice, if the plate was quickly moved to the bench, or if a cold lysis solution was added).
4. Warm lysis solution to room temperature before adding to cells.
5. Allow the lysis reaction to proceed for 8 minutes at 25 degrees C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link