Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

TaqMan™ Gene Expression Cells-to-CT™ Kit

Catalog number: AM1728
Have Questions?
Invitrogen™

TaqMan™ Gene Expression Cells-to-CT™ Kit

Catalog number: AM1728
Have Questions?
Catalog Number
AM1728
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
400 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
40 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM1728100 reactions
-
AM1729400 reactions
-
439900240 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The TaqMan™ Gene Expression Cells-to-CT™ Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification. This kit saves time and offers a simple workflow that is suitable for a few samples or can be easily incorporated into automated, high-throughput applications.

The TaqMan™ Gene Expression Cells-to-CT Kit is:
Complete—optimized workflow includes cell lysis reagents with gDNA removal, RT enzyme mix, buffer, and new TaqMan™ Gene Expression Master Mix
Fast—7-minute sample prep, including DNase treatment, at room temperature
Easy—lyse samples in a tube or directly in culture plates
Robust—perform gene expression analysis on 10–100,000 cells per sample; results equivalent to those from purified RNA
Efficient—contains sufficient reagents to generate 500 real-time PCR results from 100 starting samples

Featuring a unique method for lysing cultured cells while removing genomic DNA and preserving RNA integrity, the TaqMan™ Gene Expression Cells-to-CT™ Kit contains reverse transcription (RT) reagents for cDNA synthesis and TaqMan™ Gene Expression Master Mix for real-time PCR analysis. TaqMan™ primer⁄probe sets are sold separately.

Simple 7-minute sample preparation: part of a complete workflow
Whether you are using plates or tubes, the TaqMan™ Gene Expression Cells-to-CT™ Kit, which uses the simple 7-minute sample preparation procedure outlined (see figure), is designed for 10–100,000 cultured cells⁄sample. Cells are washed in PBS and lysed in solution for 5 minutes at room temperature; DNase treatment can be performed simultaneously. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution. The lysates are now ready for reverse transcription or storage at –20°C. Because samples can be processed directly in culture wells (96 or 384 wells), sample handling and the potential for sample loss or transfer error are minimized, facilitating rapid, high-throughput processing. Unlike old-fashioned, multi-step RNA isolation protocols, no heating, washing, or centrifugation steps are required. The kit greatly simplifies a laborious 30–60 minute process and reduces it to 7 minutes.

The TaqMan™ Gene Expression Cells-to-CT™ Kit workflow enables unsurpassed gene expression evaluation with any of the >700,000 TaqMan™ Gene Expression Assays. This kit has been extensively tested for specificity with a broad selection of TaqMan™ Gene Expression Assays and shows performance equivalent to that obtained with purified RNA (see figure).

Achieve unsurpassed performance and sensitivity
Unlike some competitor kits that limit the amount of lysate in the RT reaction to 5%, the TaqMan™ Gene Expression Cells-to-CT™ Kit can accommodate 45% of the total RT reaction volume as cell lysate. Additionally, cDNA can comprise up to 45% of the real-time PCR reaction volume. The large lysate volume in the optimized RT reaction, along with the large cDNA volume in the subsequent real-time PCR using the TaqMan™ Gene Expression Master Mix, lead to maximum sensitivity. The master mix amplifies the target precisely and accurately, enabling the detection of small quantities of target, such as transcripts expressed at low levels.

The ability to detect limited target quantities was tested by mixing a constant number of human cells with various numbers of mouse cells. The cell mixtures were prepared using the TaqMan™ Gene Expression Cells-to-CT™ and assayed for a mouse-specific and human-specific gene. Comparative data were generated in the same manner with RNA purified by traditional methodology. The data show that RNA from as few as 10 mouse cells was detectable in a background of RNA from 10,000 human cells (see figure). The ability of the TaqMan™ Gene Expression Cells-to-CT™ Kit to detect relative low abundance transcripts was equivalent to that of purified RNA; at low levels, it was superior.

Additionally, the performance of the TaqMan™ Gene Expression Cells-to-CT™ Kit was compared to competitor lysate kits and to purified RNA. Inputs of 100–100,000 cells⁄lysis reaction were examined. The sensitivity of the TaqMan™ Cells-to-CT™ Kit protocol was equivalent to that obtained with purified RNA, and it surpassed competitor lysates (see figure). Furthermore, the lack of inhibition at high cellular inputs and the low variation among technical replicates demonstrate the reliability of this approach for gene expression studies using cultured cells.

Proven performance, proven together
All components of the TaqMan™ Gene Expression Cells-to-CT™ Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate sample preparation, RT, and real-time PCR kits. And the TaqMan™ Gene Expression Cells-to-CT™ Kit has been validated with TaqMan™ Gene Expression Assays for added quality assurance.

Accessory product
The TaqMan™ Cells-to-CT™ Control Kit was designed for use with the TaqMan™ Gene Expression Cells-to-CT™ Kit. This kit contains an endogenous control (ACTB) to normalize for sample loading variability and an artifical XenoRNA™ control and corresponding TaqMan™ Gene Expression Assay to monitor the effects of PCR inhibition.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
2-step RT-qPCR
Polymerase
DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7500 System, 7900HT System, Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Green Features
Fewer resources used and less waste, Less hazardous
Passive Reference Dye
ROX (Pre-mixed)
Product Line
Ambion™, Cells-to-CT™, TaqMan™
Product Type
Gene Expression Cells-to-C Kit
Purity or Quality Grade
UP (Ultra Pure)
Reverse Transcriptase
M-MLV
Shipping Condition
Dry Ice
Sufficient For
100 Reactions
No. of Reactions
100 Reactions
Reagent Type
Cell Lysis
Sample Type
Cell

Contents & Storage

Contains:
• Stop Solution: 500 μL tube
• DNase I: 55 μL tube
• 20X RT Enzyme Mix: 275 μL tube
• Lysis Solution: 5.5 mL bottle
• 2X RT Buffer: 5.5 mL bottle
• TaqMan™ Gene Expression Master Mix: 5 m bottle

Store Stop Solution, DNase I, and 20X RT Enzyme Mix at -20°C.
Store Lysis Solution, 2X RT Buffer, and TaqMan™ Gene Expression Master Mix at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text