Invitrogen™

ArrayControl™ RNA Spikes

Catalog number: AM1780
Have Questions?
Invitrogen™

ArrayControl™ RNA Spikes

Catalog number: AM1780
Have Questions?
Catalog Number
AM1780
Unit Size
8 tubes
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM17808 tubes
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Ambion™ ArrayControl™ RNA Spikes consist of 8 individual RNA transcripts ranging in size from 750 to 2,000 bases with a 30-base 3' poly(A) tail. RNA Spikes are added to the RNA sample and used as template for preparing labeled cDNA. Each transcript is supplied in one tube containing 1 μg.
• For the normalization and control of microarray experiments
• For array printing and protocol assessment/optimization
• Monitor reverse transcription and cDNA labeling efficiency, hybridization stringency, and background
• Normalize Cy™3 and Cy™5 signals When hybridized to complementary spots on a gene array, ArrayControl™ RNA Spikes can be used to monitor the efficiency of reverse transcription and aRNA amplification, labeling, and hybridization. They can also be used to assess the precision, accuracy, and consistency of array printing and protocol sensitivity. In addition, RNA Spikes are an invaluable tool for protocol optimization and troubleshooting. They can be used for normalization of Cy™3 and Cy™5 signals, determination of dynamic range, and slide orientation. Individual RNA Spikes can also be omitted to serve as a negative control for background calculations. The RNA Spike sequences have been carefully chosen to lack homology to mammalian sequences that might otherwise lead to cross-hybridization to gene-specific spots. Each RNA Spike is purified, precisely quantitated by spectrophotometry and RiboGreen™ staining, and tested for stability under highly stringent conditions. They are also tested for cDNA labeling efficiency using both direct and indirect methods. Their manufacturing and quantitation are consistent from lot-to-lot, so that signals can be compared between experiments and to assess array reproducibility. To obtain RNA Spike sequence information for the preparation of complementary control spots on your arrays, please contact Technical Support.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
RNA Spike
Label or Dye
Unconjugated
Shipping Condition
Dry Ice
Product Line
Ambion™, ArrayControl™
Quantity
8 Tubes

Contents & Storage

RNA Spikes 1–8 should be stored below –70°C. TE Buffer should be stored at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text