MagMAX™ Total Nucleic Acid Isolation Kit
MagMAX™ Total Nucleic Acid Isolation Kit
Applied Biosystems™

MagMAX™ Total Nucleic Acid Isolation Kit

The MagMAX™ Total Nucleic Acid Isolation Kit is designed for the rapid, high-throuphput purification of RNA and genomic DNA fromRead more
Have Questions?
Catalog NumberQuantity
AM1840100 preps
Catalog number AM1840
Price (EUR)
1.040,00
Each
Add to cart
Quantity:
100 preps
Price (EUR)
1.040,00
Each
Add to cart
Ask our AI about this Product
The MagMAX™ Total Nucleic Acid Isolation Kit is designed for the rapid, high-throuphput purification of RNA and genomic DNA from a variety of samples including viral, blood, and bacterial samples. It includes zirconia beads for the mechanical disruption of difficult pathogens. RNA purified using the kit is ideal for PCR and RT-PCR applications. The kit uses magnetic bead–based purification and includes sufficient reagents for 100 reactions.

• Complete, ready-to-use reagents for RNA and DNA purification from tough-to-lyse bacteria
• Highly effective sample homogenization using bead beating and chemical lysis
• Magnetic bead–based purification technology allows effective removal of PCR inhibitors
• Nucleic acid purification is easily scalable and suitable for automation

Proven with a Wide Range of Samples:
• Biological Samples
• Feces and whole blood
• Biofluids such as milk, urine, nasal fluids, swab samples, semen, sputum, and spent culture medium
• Bacterial cultures and mammalian cell cultures
• Environmental Samples
• River and pond water
• Agricultural samples such as manure pits and cow alleyways

Benefits of the MagMAX™ Total Nucleic Acid Isolation Kit:
The MagMAX™ Total Nucleic Acid Isolation Kit procedure combines an effective method sample disruption with magnetic bead–based nucleic acid isolation and purification technology. The MagMAX™ microspherical paramagnetic beads have a large available binding surface and can be fully dispersed in solution, allowing thorough nucleic acid binding, washing, and elution. This results in consistent yields of high-quality nucleic acid with >50% recovery and little sample-to-sample variation (recovery may vary depending on sample type). The entire procedure can be completed in approximately 1 hour.

Overview of the Procedure:
Samples are disrupted using zirconia beads in a guanidinium thiocyanate–based solution; this rapidly releases nucleic acid while simultaneously inactivating nucleases. Samples are then diluted with isopropanol, and paramagnetic beads are added to the sample. The beads bind nucleic acids and are then immobilized on magnets and washed to remove proteins and other contaminants. A second wash solution is used to remove residual binding solution. Finally, nucleic acid is eluted in a small volume of low-salt buffer. The MagMAX™ Total Nucleic Acid Isolation Kit procedure is designed for processing samples in 96-well plates, thus 96 samples can be processed at once; however, the kit can also be used with fewer than 96 samples.

Accessory Products:
Lysis/Binding Solution Concentrate (SKU#AM8500), Wash Solution 1 Concentrate (SKU#AM8504), and Wash Solution 2 Concentrate (SKU#AM8640) are components of the MagMAX™ Total Nucleic Acid Isolation Kit that may also be purchased separately.
For Veterinary Use Only.
Specifications
Final Product TypeRNA, DNA
For Use With (Application)Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Southern Blotting, PCR, Northern Blotting
High-throughput CompatibilityHigh-throughput Compatible
Product LineMagMAX™
Quantity100 preps
Sample TypeBacteria, Blood, Cells, Liquid Samples (e.g. Serum)
Starting Material AmountUp to 0.3 g solid samples or 175 μl liquid samples
TargetTotal RNA, Total Nucleic Acids, Genomic DNA
Test Time1 hr.
Yield420 ng
Format96-well Plate
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage

• Carrier NA and Lysis/Binding Enhancer, store at -20°C
• RNA Binding Beads, 4°C
• Wash Solution 2 Concentrate, 4°C or room temperature
• Elution Buffer, 4°C or room temperature
• Lysis/Binding Solution Concentrate, room temperature
• Wash Solution 1 Concentrate, room temperature
• Processing Plates, room temperature
• Lids, room temperature
• PBS Buffer, room temperature

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
00871302Certificate of AnalysisFeb 07, 2020AM1840
00867239Certificate of AnalysisJan 31, 2020AM1840
00866083Certificate of AnalysisJan 23, 2020AM1840
00866158Certificate of AnalysisJan 23, 2020AM1840
00865978Certificate of AnalysisJan 23, 2020AM1840
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link