SUPERase•In RNase Inhibitor is a protein-based inhibitor of non-human origin that noncovalently binds and inhibits the most common and troublesomeRead more
Have Questions?
Change view
Catalog Number
Quantity
AM2696
10,000 U
AM2694
2500 U
2 Options
Catalog number AM2696
Price (USD)
604.00
Each
In stock
Add to cart
Quantity:
10,000 U
Request bulk or custom format
Price (USD)
604.00
Each
Add to cart
SUPERase•In RNase Inhibitor is a protein-based inhibitor of non-human origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1.
SUPERase•In RNase Inhibitor can be used in any application where RNase contamination could be problematic. Because it inhibits a broader range of RNases than traditional RNase inhibitors, SUPERase•In is the most effective RNase inhibitor available, providing a higher level of protection against degradation.
SUPERase•In does not interfere with other enzymes such as RNA polymerases, reverse transcriptase, or Taq DNA polymerase. Additionally, SUPERase•In is active up to 65°C and over a pH range of 5.5 to 8.5.
If you are plannning to use this product with standard antibody purification methods, please contact our technical support to discuss experimental design.
Unit definition SUPERase•In RNase Inhibitor at 1 U/μL will block the degradation of 0.1 μg/μL labeled RNA by 2.5 pg/μL of RNase A, 2.5 pg/μL of RNase I, and 0.0075 U/μL of RNase T1, for 4 hours at 37°C, in 20 mM Tris-HCl, pH 7.5, 50 mM NaCl, 1 mM EDTA. Analysis is by denaturing PAGE. SUPERase•In is currently the only ribonuclease inhibitor for which the unit activity is defined by such a functional assay.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration50 mM
Product LineAmbion™, SUPERase•In™
Quantity10,000 U
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Supplied in a single tube that shouldbe stored at -20°C.
Storage buffer: 2 mM KH2PO4, 8 mM Na2HPO4,2.7 mM KCl, 137 mM NaCl, pH 7.4 in 50% glycerol
Frequently asked questions (FAQs)
What temperature and pH is SUPERase·In RNase Inhibitor (Cat. No. AM2694, AM2696) active up to?
SUPERase·In RNase Inhibitor (20 U/µL) (Cat. No. AM2694, AM2696) is active up to 65 degrees C and over a pH range of 5.5 to 8.5.
What kind of Rnases can be inhibited using SUPERase.In RNase Inhibitor?
SUPERase.In RNase Inhibitor is a protein-based inhibitor of nonhuman origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1.