This high-purity Ambion™ enzyme is a mix of cloned RNA polymerase and RNase inhibitor. It has a high specificity forRead more
Have Questions?
Catalog Number
Quantity
AM2718
30000 units, 200 μL
Catalog number AM2718
Price (USD)
658.65
Online Exclusive
706.00
Save 47.35 (7%)
Each
Add to cart
Quantity:
30000 units, 200 μL
Request bulk or custom format
Price (USD)
658.65
Online Exclusive
706.00
Save 47.35 (7%)
Each
Add to cart
This high-purity Ambion™ enzyme is a mix of cloned RNA polymerase and RNase inhibitor. It has a high specificity for its promoter, transcribing large amounts of RNA with no cross talk between promoters. It is supplied in one tube containing 30,000 U at a concentration of 200 U/ μL. The enzyme incorporates radiolabeled nucleotides along with many modified nucleotides. Inclusion of RNase inhibitor provides broad-spectrum protection against enzymatic degradation of the synthesized RNA, making the enzyme ideal for the synthesis of RNA for blot and in situ hybridization probes, RPA analysis, in vitro translation, and antisense RNA.
Unit Definition: One unit is the amount of T7 RNA Polymerase-Plus Enzyme Mix required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration200 U/μL
Quantity30000 units, 200 μL
Shipping ConditionDry Ice
SourceDNA
PolymeraseT7 RNA Polymerase
Product LineAmbion™
PromoterT7
Unit SizeEach
Contents & Storage
Store at –20°C.
Please enter sku to load default questions
Frequently asked questions (FAQs)
Can I substitute the enzyme mix in the MEGAscript T7 Transcription Kit (Cat. No. AM1333) with T7 RNA Polymerase-Plus Enzyme Mix (Cat. No. AM2718)?
No, we do not recommend substituting the enzyme mix in the MEGAscript T7 Transcription Kit (Cat. No. AM1333) with T7 RNA Polymerase-Plus Enzyme Mix (Cat. No. AM2718).