Ambion™ Silencer™ GAPDH Positive Control siRNA (human) is ideal for developing and optimizing siRNA experiments. The control is validated for use in human cell lines. 5 nmol is provided, along with an additional tube of 2 nmol negative control siRNA. • Validated siRNA controls for optimizing siRNA experiments • Gene-specific control siRNAs provided with scrambled, negative controls • Functionally tested in several common cell lines • HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Ambion™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format
Tube
Control Type
Positive Control
RNAi Type
siRNA
Species
Human
Sample Type
Cell Cultures
For Use With (Application)
RNAi
Label or Dye
Unconjugated
Product Line
Silencer™, Ambion™
Product Type
siRNA
Purity
HPLC
Quantity
40 nmol
Shipping Condition
Room Temperature
Target
GAPDH
Contents & Storage
The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.