Yeast RNA (10 mg/mL)
Inquire about OEM or Commercial Supply version of this product here.
Yeast RNA (10 mg/mL)
Invitrogen™

Yeast RNA (10 mg/mL)

Purified Torulla Ambion™ yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactionsRead more
Have Questions?
Catalog number AM7118
Price (USD)/ 10 tubes
363.65
Online exclusive
368.00 
Save 4.35 (1%)
-
Add to cart
Price (USD)/ 10 tubes
363.65
Online exclusive
368.00 
Save 4.35 (1%)
Add to cart
Yeast RNA (10 mg/mL)
Catalog numberAM7118
Price (USD)/ 10 tubes
363.65
Online exclusive
368.00 
Save 4.35 (1%)
-
Add to cart
Purified Torulla Ambion™ yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant. This total RNA preparation is supplied in ten 1 mL tubes at a concentration of 10 mg/mL in diethyl pyrocarbonate-treated distilled water. It is typically used at a working concentration of 10–20 μg/mL.

What is a Coprecipitant?
Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration10 mg/mL
For Use With (Application)Carrier for Nucleic Acid Precipitation
Product TypeYeast RNA
Product LineAmbion™
Quantity10 x 10 mg
Shipping ConditionDry Ice
Unit Size10 tubes
Contents & Storage
Store at –20°C.