1x1 image pixel for data collection
Century™-Plus RNA Markers
Century™-Plus RNA Markers
Invitrogen™

Century™-Plus RNA Markers

Ambion™ RNA Century™-Plus Markers are a set of 7 in vitro transcripts of 100, 200, 300, 400, 500, 750, andRead more
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Catalog NumberQuantity
AM7145Promo Image1 tube
Catalog number AM7145
Price (USD)
291.00
Each
In stock
Add to cart
Quantity:
1 tube
Request bulk or custom format
Price (USD)
291.00
Each
Add to cart
Ambion™ RNA Century™-Plus Markers are a set of 7 in vitro transcripts of 100, 200, 300, 400, 500, 750, and 1000 bases. They are unlabeled for staining with EtBr and are provided in one tube containing 50 μg at a concentration of 1 mg/mL. They are premixed to give equivalent staining on a 5% polyacrylamide/8 M urea gel and are useful as RNA size standards in denaturing gel electrophoresis. 1–2 μg (2–4 μL) markers will generate 7 distinct bands on a TBE-Urea acrylamide gel with EtBr staining. Markers are also tested to be nuclease-free and stable overnight at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mg/ml
Gel CompatibilityDenaturing Agarose Gels, Denaturing Polyacrylamide Gels
Mass50 μg
Product LineAmbion™, RNA Century™
Product TypeRNA Marker
Quantity1 tube
Ready to LoadNo
Shipping ConditionDry Ice
Size Range0.1 to 1 kb
Unit SizeEach
Contents & Storage
Store below –70°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection