Invitrogen™

Human Heart Total RNA

Catalog number: AM7966
Have Questions?
Invitrogen™

Human Heart Total RNA

Catalog number: AM7966
Have Questions?
Catalog Number
AM7966
Unit Size
100 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM7966100 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The highest-quality RNA available from Ambion®, it is DNase-treated, subjected to unsurpassed quality control standards, and certified to contain small RNAs (miRNA, siRNA, and snRNA). One tube containing 100 μg is provided at a concentration of 1 mg/mL. Intactness of mRNA, genomic DNA contamination, presence/absence of enzymatic inhibitors, and ribosomal RNA contamination in poly(A) preparations are all important aspects used to judge the quality of FirstChoice® RNAs. The samples are processed using Ambion® RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Note: While it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent® 2100 bioanalyzer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
1 mg/mL
Species
Human
Sample Type
Total RNA
Product Line
Ambion™
Purity
microRNA-Certified
Sample Treatment
DNase-Treated
Sample Source
Normal Tissue
Shipping Condition
Dry Ice
Tissue
Heart

Contents & Storage

Store at or below -70°C. Avoid multiple freeze–thaw cycles. Aliquots of the product may be stored short-term at -20°C. DO NOT STORE IN FROST-FREE FREEZER.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text