Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

5-(3-Aminoallyl)-UTP (50 mM)

Catalog number: AM8437
Have Questions?
Invitrogen™

5-(3-Aminoallyl)-UTP (50 mM)

Catalog number: AM8437
Have Questions?
Catalog Number
AM8437
Unit Size
50 µl
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM843750 µl
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Ambion modified nucleotides confer unique characteristics to the RNA molecules into which they are incorporated. Incorporation of an aminoallyl-modified NTP, for example, allows for the subsequent coupling of the reactive amino group to another molecule such as biotin or a Cy dye. Unlike NTPs directly coupled to a dye, aminoallyl-modified NTPs are incorporated almost as efficiently as unmodified NTPs and are much less expensive than dye-coupled NTPs.

Ambion 5-(3-Aminoallyl)-UTP (50 mM) is supplied in a single tube at 50 mM.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Nucleotides
Shipping Condition
Dry Ice
Concentration
50 mM
Label or Dye
Aminoallyl
Quantity
50 μL
Product Line
Ambion™
For Use With (Application)
Nucleic Acid Labeling
Form
Liquid

Contents & Storage

5-(3-Aminoallyl)-UTP, 50 mM (50 μL)

Store at –70°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text