1x1 image pixel for data collection
EDTA (0.5 M), pH 8.0, RNase-free
EDTA (0.5 M), pH 8.0, RNase-free
Invitrogen™

EDTA (0.5 M), pH 8.0, RNase-free

Ambion Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle containing 100 mL. The solutionRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM9260G100 mL
AM9261500 mL
AM92621 L
Catalog number AM9260G
Price (USD)
58.65
Online Exclusive
61.75
Save 3.10 (5%)
Each
In stock
Add to cart
Quantity:
100 mL
Price (USD)
58.65
Online Exclusive
61.75
Save 3.10 (5%)
Each
Add to cart

Ambion Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Ambion's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialEDTA
Packaging TypeBottle
Product LineAmbion™
PurityMolecular Biology Grade
Quantity100 mL
Shipping ConditionRoom Temperature
Concentration0.5 M
FormLiquid
pH8
Unit SizeEach
Contents & Storage
Store at room temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link