KCl (2 M), RNase-free
KCl (2 M), RNase-free
Invitrogen™

KCl (2 M), RNase-free

Ambion® Molecular biology grade, 2 M KCl solution is supplied in one bottle containing 100 mL. The solution is certifiedRead more
Have Questions?
Catalog NumberQuantity
AM9640G100 mL
Catalog number AM9640G
Price (USD)
54.65
Online Exclusive
58.00
Save 3.35 (6%)
Each
-
Add to cart
Quantity:
100 mL
Price (USD)
54.65
Online Exclusive
58.00
Save 3.35 (6%)
Each
Add to cart
Ask our AI about this Product
Ambion® Molecular biology grade, 2 M KCl solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Ambion®'s nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPotassium Chloride
For Use With (Application)RNA Applications
Packaging TypeBottle
Product LineAmbion™
PurityMolecular Biology Grade
Quantity100 mL
Shipping ConditionRoom Temperature
Concentration2 M
FormLiquid
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link