1x1 image pixel for data collection
RNA Rapid Extraction Solution
RNA Rapid Extraction Solution
Invitrogen™

RNA Rapid Extraction Solution

Ambion® Whatman® FTA® Cards are frequently used for the collection, transport, and storage of RNA and DNA in biological samples.Read more
Have Questions?
Catalog NumberQuantity
AM977510 mL
Catalog number AM9775
Price (USD)
182.00
Each
Add to cart
Quantity:
10 mL
Price (USD)
182.00
Each
Add to cart
Ambion® Whatman® FTA® Cards are frequently used for the collection, transport, and storage of RNA and DNA in biological samples. The RNA Rapid Extraction Solution is to release RNA from the paper prior to RNA isolation. 10 mL is provided.
• Rapid 5 min procedure to release RNA from Whatman® FTA® cards
• Ideal for high-throughput applications
• Integrity of the RNA is unaffected
• Ready for real-time PCR Disks punched from FTA® Cards are simply dropped into RNA Rapid Extraction Solution and agitated for 5 minutes to release the sample for RNA isolation—while protecting the RNA from degradation. The RNA can then be purified from the RNA Rapid Extraction Solution using the MagMAX™-96 Viral RNA Isolation Kit (SKU #AM1836). The high-quality purified RNA is suitable for quantitative RT-PCR or other downstream RNA applications. Whatman® FTA® Cards can be used with virtually any sample type including blood, cultured cells, buccal cells, plant material, bacteria, plasmids, microorganisms, solid tissue, viral particles, and more.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeTotal RNA, Viral DNA/RNA
For Use With (Application)Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
For Use With (Equipment)MagMAX™-96 Viral RNA Isolation Kit
High-throughput CompatibilityNot High-throughput Compatible (Manual)
IncludesSolution, without manual
Product LineAmbion™
Quantity10 mL
Shipping ConditionRoom Temperature
Isolation TechnologyOrganic Extraction
Sample TypePlant Samples, Blood, Other Samples, Bacteria, Cells, Whatman FTA Cards Samples
TargetViral DNA/RNA, Total RNA
Unit SizeEach
Contents & Storage
Store at room temperature.

Share catalog number, name or link