Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

RNaseZap™ RNase Decontamination Solution

Catalog number: AM9780
Have Questions?
Invitrogen™

RNaseZap™ RNase Decontamination Solution

Catalog number: AM9780
Have Questions?
Catalog Number
AM9780
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 250 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM9780250 mL
-
AM97826 x 250 mL
-
AM97844 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

RNaseZap™ RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap™ Solution onto the surface to be decontaminated and rinse it off with RNase-free water.

Features of RNaseZap™ RNase Decontamination Solution:
• Completely removes RNase contamination from glass and plastic surfaces
• Excels at removing high levels of RNase contamination whereas similar products fail
• Proven effective at removing high concentrations of dried-on RNase A
• Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free

Using RNaseZap™ Solution
Working with RNA requires that special measures be taken to ensure an RNase-free environment. Even trace quantities of RNase can lead to lower yields from in vitro transcription reactions, degradation during RNA purification protocols, and variable results in RPAs and Northerns. RNaseZap™ contains three ingredients active against RNase and has proven to be extremely effective at removing RNase contamination from glassware, plastic surfaces, countertops, and pipettors. It has also been shown to be effective at eliminating RNase contamination from microfuge tubes without inhibiting subsequent enzymatic reactions. Supplied in one 250 mL bottle.

RNase contamination in microfuge tubes
While concern about RNase contamination of frequently handled labware is common and prudent, researchers often do not worry about RNase contamination in commercially prepared products such as microfuge tubes. However, in a survey of six standard 1.5 mL microfuge tubes in which three were pre-rinsed with RNaseZap™ and water and three were left untreated, all of the untreated tubes had at least marginal levels of RNase contamination, whereas none of the tubes pre-rinsed with RNaseZap™ exhibited contamination. Additionally, in testing commercially available “RNase-free” microfuge tubes, approximately 10% had some RNase contamination (data not shown). This may well account for the erratic results that researchers periodically experience when performing RNA-based assays.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Surface decontamination solution
Shipping Condition
Room Temperature
Quantity
250 mL
Reagent Type
RNase Control Reagent
Product Line
Ambion™, RNaseZap

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text