TE, pH 8.0, RNase-free
TE, pH 8.0, RNase-free
Invitrogen™
TE, pH 8.0, RNase-free
Ambion molecular biology grade, TE, pH 8.0 solution is supplied in one bottle containing 500 mL. The buffer is certifiedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM98581 L
AM9849500 mL
Catalog number AM9858
Price (USD)
93.65
Online Exclusive
101.00
Save 7.35 (7%)
Each
Add to cart
Quantity:
1 L
Price (USD)
93.65
Online Exclusive
101.00
Save 7.35 (7%)
Each
Add to cart
Ambion molecular biology grade, TE, pH 8.0 solution is supplied in one bottle containing 500 mL. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Ambion's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Recommended StorageStore at room temperature.
Physical FormLiquid
Product LineAmbion
Quantity1 L
pH8.0
Unit SizeEach
Share catalog number, name or link