Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

TAE (10X), RNase-free

Catalog number: AM9869
Have Questions?
Invitrogen™

TAE (10X), RNase-free

Catalog number: AM9869
Have Questions?
Catalog Number
AM9869
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM98691 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Molecular biology grade, Ambion® 10X TAE is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Ambion®'s nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name or Material
Running Buffer
Recommended Storage
Store at room temperature.
Concentration
10X
For Use With (Application)
Nucleic acid gel electrophoresis & blotting
Physical Form
Liquid
Product Line
Ambion
Quantity
1 L
Grade
Molecular Biology

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text