EDTA (0.1 mM), pH 8.0, RNase-free
Inquire about OEM or Commercial Supply version of this product here.
EDTA (0.1 mM), pH 8.0, RNase-free
Invitrogen™

EDTA (0.1 mM), pH 8.0, RNase-free

Ambion® 0.1 mM EDTA is a commonly recommended storage solution for RNA samples. 0.1 mM EDTA is in nuclease-free, ultrapureRead more
Have Questions?
Catalog number AM9912
Price (USD)/ 50 mL
63.65
Online exclusive
65.25 
Save 1.60 (2%)
-
Add to cart
Price (USD)/ 50 mL
63.65
Online exclusive
65.25 
Save 1.60 (2%)
Add to cart
EDTA (0.1 mM), pH 8.0, RNase-free
Catalog numberAM9912
Price (USD)/ 50 mL
63.65
Online exclusive
65.25 
Save 1.60 (2%)
-
Add to cart
Ambion® 0.1 mM EDTA is a commonly recommended storage solution for RNA samples. 0.1 mM EDTA is in nuclease-free, ultrapure water. It is provided in one bottle of 50 mL. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. 0.1 mM EDTA is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialEDTA
Concentration0.1 M
Sample TypeRNA
For Use With (Application)Nucleic Acid Gel Electrophoresis & Blotting
Packaging TypeBottle
pH8
Product LineAmbion™
PurityDNase-Free, RNase-Free
Quantity50 mL
Shipping ConditionRoom Temperature
FormLiquid
Unit Size50 mL
Contents & Storage
Store at room temperature.