Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Nuclease-Free Water (not DEPC-Treated)

Catalog number: AM9937
Have Questions?
Invitrogen™

Nuclease-Free Water (not DEPC-Treated)

Catalog number: AM9937
Have Questions?
Catalog Number
AM9937
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 1,000 ml
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 1,000 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM993710 x 50 mL
-
AM99381 x 100 mL
-
AM99301 x 500 mL
-
AM99395 x 100 mL
-
AM99321 x 1,000 ml
-
43879364 x 1,000 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving. This nuclease-free water is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Using Nuclease-free Water
Because this water is not DEPC-treated, it is recommended for applications for which residual DEPC might prove detrimental (e.g., oocyte microinjection).

For Research Use Only. Not for use in diagnostic procedures.

Specifications

pH
6 to 8
Recommended Storage
Store at room temperature.
Grade
Molecular Biology
Packaging
Bottle
Treatment(s)
Not DEPC-Treated
Quantity
10 x 50 mL
Purification Method
Autoclaved, Membrane-Filtered
Shipping Condition
Room Temperature
Product Line
Ambion™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text