Applied Biosystems™

MagMAX™-96 Viral RNA Isolation Kit

Catalog number: AMB18365
Have Questions?
Applied Biosystems™

MagMAX™-96 Viral RNA Isolation Kit

Catalog number: AMB18365
Have Questions?
Catalog Number
AMB18365
Unit Size
5 x 96 preps
Price (USD)
Quantity
Catalog Number
Unit Size
96 preps
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AMB183655 x 96 preps
AM183696 preps
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The MagMAX-96 Viral RNA Isolation Kit is for the isolation of viral RNA and DNA from diverse sample types (e.g., environmental samples and biofluids). Nucleic acids purified using this kit are ideal for PCR and RT-PCR applications. For a NVSL-validated version of this kit, please see the MagMAX-96 AI⁄ND Viral RNA Isolation Kit.

Benefits of MagMAX magnetic bead-based purification:
Magnetic beads offer many benefits compared to other technologies for isolating RNA. Beads bind RNA more efficiently than glass fiber filters, resulting in higher and more consistent RNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high-efficiency binding, the bound RNA can be eluted in just 20–50 μL of nuclease-free water, concentrating RNA from large, dilute samples.

Optimized for isolation of virus from biofluids
The MagMAX Viral RNA Isolation Kit has been optimized for use with biological fluids and cell-free samples such as serum, plasma, swabs, and cell culture media. These samples often present unique challenges when purifying RNA, including low viral titers, large volumes to be processed, and sample complexity. Without the need for organic solvents or RNA precipitation, the MagMAX Viral RNA Isolation Kit eliminates the problems often encountered with other RNA isolation methods such filter clogging, large elution volumes, and inconsistent yields. With this kit, DNA and RNA input levels ranging from 20 copies to 25 x 106 copies have been quantitatively recovered from plasma, serum, and milk. And even with less sample input, pathogen detection was as sensitive (equivalent CTs) or more sensitive (lower CTs) using nucleic acid isolated with the MagMAX kit when compared to nucleic acid isolated using another company's product.

Accessory Products:
This kit is optimized for automated nucleic acid purification using KingFisher magnetic particle processors. This kit can also be used for manual nucleic acid purification using a 96-well magnetic ring stand. The Magnetic Stand-96 (Cat. No. AM10027) and the 96-well Magnetic Ring Stand (Cat. No. AM10050) are designed for paramagnetic bead precipitation from standard 96-well, U-bottom microplates. The Magnetic-Ring Stand pellets the magnetic beads in a circle shape, enabling easy removal of supernatant by a robot or multichannel pipettors.

Specifications

Format
96-well Plate
Isolation Technology
Magnetic Bead
Purification Time
45 min.
Sample Type
Cell-free Biological Fluids, Liquid Samples (e.g. Serum), Viral Samples
Final Product Type
Viral DNA⁄RNA
High-throughput Compatibility
High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
For Use With (Equipment)
KingFisher™ Flex Magnetic Particle Processor, KingFisher 96
For Use With (Application)
Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
No. of Reactions
480
Product Line
MagMAX™
Quantity
5 x 96 Preps
Scale
Maxi
Starting Material Amount
Up to 50 μl
Target
Viral DNA/RNA
Yield
5 ng

Contents & Storage

Carrier RNA and Lysis Binding Enhancer is stored at –20°C; RNA Binding Beads are stored at 4°C. Wash Solution 2 Concentrate and Elution Buffer may be stored at either 4°C or room temperature. Lysis⁄Binding Solution Concentrate, Wash Solution 1 Concentrate, and Processing Plates and Lids are stored at room temperature. Nuclease-free water may be stored at any temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text