EVOS™ Arm Rest Kit
Catalog number: AMEP4618
Have Questions?
EVOS™ Arm Rest Kit
Catalog number: AMEP4618
Have Questions?
Catalog Number
AMEP4618
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AMEP46181 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The EVOS Arm Rest is designed to help improve precision and reduce fatigue during extended operations, such as cell isolation and tissue culture. It fits on either side of the instrument stage for optimal positioning, and the non-skid surface helps prevent slipping. The EVOS Arm Rest Kit was designed for use with the EVOS XL, FL, and FL Color imaging systems.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Type
Arm Rest Kit
Product Line
EVOS
For Use With (Equipment)
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System
Quantity
1 Ea.