Thermo Scientific™
EVOS™ Power Cord, Type G (United Kingdom)
Catalog number: AMEP4645
Have Questions?
Thermo Scientific™
EVOS™ Power Cord, Type G (United Kingdom)
Catalog number: AMEP4645
Have Questions?
Catalog Number
AMEP4645
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AMEP46451 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries or regions with Type G power plugs. The Type G electrical plug is used in the UK, Ireland, Hong Kong, Singapore, and other parts of Africa and Asia.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Type
Power Cord
Product Line
EVOS
Connector Types
Type G (Electrical Plug), Type G (Power Plug)
For Use With (Equipment)
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Quantity
1 Ea.