Informational:Informational:Experience nuclei counting with fluorescent-mode counting algorithms, simplified workflows, built-in dilution calculators and reports using the latest version of Countess software with optional 21 CFR Part 11 compliance. Download now!
Thermo Scientific™

Countess™ Disposable Slide Holder

Catalog number: AMEP4745
Have Questions?
Thermo Scientific™

Countess™ Disposable Slide Holder

Catalog number: AMEP4745
Have Questions?
Catalog Number
AMEP4745
Unit Size
1 unit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AMEP47451 unit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
For use with Countess II FL, Countess 3, and Countess 3 FL automated cell counters. Holds Countess disposable slides. Not compatible with the Countess™ II FL Reusable Slide (Cat. No. A25750).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Disposable Slide Holder
Product Line
Countess™
For Use With (Application)
Cell Counting
For Use With (Equipment)
Originally supplied with CIIFL Automated Counter
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text