Invitrogen™

EVOS™ Onstage Vessel Holder, two Lab-Tek or Lab-Tek-II coverglass chamber slides

Catalog number: AMEPVH069
Have Questions?
Invitrogen™

EVOS™ Onstage Vessel Holder, two Lab-Tek or Lab-Tek-II coverglass chamber slides

Catalog number: AMEPVH069
Have Questions?
Catalog Number
AMEPVH069
Unit Size
1 holder
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AMEPVH0691 holder
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
EVOS Onstage vessel holders maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FL Auto Imaging System equipped with an EVOS Onstage Incubator. EVOS Onstage vessel holders allow a perfect fit of your micro-well plates, cell culture flasks/dishes, and slides to the master plate of the EVOS Onstage Incubator for increased precision in sample alignment. This particular vessel holder accommodates two Nunc Lab-Tek or Lab-Tek-II coverglass chamber slides. With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy.

Learn more about the EVOS FL Auto Imaging System >

Explore the entire EVOS line of imaging systems and accessories >
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Imaging System Vessel Holder
Product Line
EVOS
Description
Onstage Vessel Holder, two Lab-Tek or Lab-Tek-II coverglass chamber slides
For Use With (Equipment)
EVOS™ FL Auto Imaging System
Holds
2 LabTech-II Chamber Slides, 2 Nunc LabTech-I
Dimensions
159 mm x 109 mm
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text