Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
AMEPVP01 | 1 each | 134.00 | - |
This EVOS vessel plate is intended for use on the stage of the EVOS FL Color Imaging System. This plate is designed to hold large vessels, for increased precision in sample alignment and is just one of many easy-to-change plates and holders available for this instrument platform. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.