AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
Actual product may vary
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
Thermo Scientific™

AquaPro™ Multi-Input Intelligent Process Transmitters

Streamline process monitoring with the Thermo Scientific™ AquaPro™ Multi-Input Intelligent Transmitter. The AquaPro provides accurate, user-friendly monitoring in-line via a reliable, simplified interface, and can be upgraded in the field to accept up to four inputs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChannel 1Voltage
AP1XXXKpH90/240 V
AP2XXXKORP100/240 V
AP2XXXJORP24 V
AP3XXXKContacting Conductivity100/240 V
AP3XXXJContacting Conductivity24 V
AP4XXXJToroidal Conductivity24 V
AP5XXXKDissolved Oxygen100/240 V
AP5XXXJDissolved Oxygen24 V
AP6XXXKDissolved Ozone100/240 V
AP6XXXJDissolved Ozone24 V
AP7XXXJRDO Pro-X optical DO24 V
AP8XXXJAquasensors Datastick24 V
AP9XXXKUltra-pure pH100/240 V
AP9XXXJUltra-pure pH24 V
AP1XXXJpH24 VDC
AP4XXXKToroidal Conductivity90/240 V
AP7XXXKRDO Pro-X optical DO90/240 V
AP8XXXKAquasensors Datastick90/240 V
Catalog number AP1XXXK
Price (USD)
1,720.00
Each
Add to cart
Channel 1:
pH
Voltage:
90/240 V
Price (USD)
1,720.00
Each
Add to cart
Streamline your operation with the Thermo Scientific™ AquaPro™ Multi-Input Intelligent Transmitter, with pre-installed sensing card. Featuring the ability to simultaneously display up to four (4) inputs or parameters, the AquaPro provides accurate, user-friendly monitoring of in-line process applications via a compact, simplified interface. From large industrial operations, to small municipal plants, the AquaPro offers the flexibility and versatility required to meet a variety of measurement needs.

Functional design

Obtain a comprehensive overview of your processes by way of a detailed picture on a single display. With the AquaPro you can reduce the time, labor and capital equipment costs associated with having multiple transmitters (installation, operation, maintenance, upgrades, etc.). Engineered to be straight-forward and intuitive, the monitor and user-interface help make your job easier. Containing a large 4.3 in. (109.22 mm) backlit color display, the AquaPro provides crystal-clear visibility in various lighting conditions. Color-coded alerts help communicate errors and warning signals, while operational menus remain simple and direct.

Unmatched versatility

Whether in the field, industrial plant, or on the manufacturing floor, the AquaPro keeps you primed and prepped to handle a variety of situations. Operators have the ability to monitor a broad range of parameters for diverse applications. Equipped with one (1) sensor card as standard, users can track up to four (4) sensors (analog or digital) by simply adding additional plug-and-play cards. Monitor pH/ORP, conductivity, dissolved oxygen, dissolved ozone, turbidity and suspended solids- the AquaPro is up for the challenge.

Purpose built

Designed to tackle the realities and utility of industrial operations, the AquaPro is built to last. Housed in a UV-resistant polycarbonate case, with NEMA 4X and IP66 waterproof ratings, you can trust that the AquaPro will remain operable in a variety of harsh industrial environments. Adaptable mounting options also provide the ability to configure the transmitter to your needs- the AquaPro is easily pipe, panel or wall-mounted.

Clear communication

Supporting most major communication protocols, the AquaPro can be used with a wide range of data systems for different applications- including information transfer to PLC, DCS, SCADA and other PC-based control systems and 0/4-20mA, MODBUS™ RTU, Ethernet/IP and Modbus TCP. Using the USB data port, data can be easily downloaded and new firmware quickly installed. This means new features and capabilities without stopping to change hardware. And in instances where additional transmitters are required, the configuration of an existing AquaPro can be easily saved and copied to a newly installed AquaPro using a removable USB drive.

Specifications
Analog OutputsTwo isolated 0/4-20mA per channel (non-reversable), 0.01mA resolution, linear/log/PID capability
Case MaterialPolycarbonate
Channel 1pH
Channels4 (Customizable with modules)
Communication DetailsOptional MODBUS RTU or Ethernet/IP
DIN Size1/2 DIN
DescriptionIntelligent multi-input transmitter with pre-installed sensing card for analog pH (combination/differential) sensors
Dimensions (L x W x H)144 x 144 x 128 mm (5.67 x 5.67 x 5.01 in.)
Display Type109.22 mm (4.3 in) high-contrast color graphical display
Electrical Requirements100 to 240VAC, 9.6W, 50/60Hz
Enclosure RatingNEMA 4X, IP66
For Use With (Equipment)Aquasensors AnalogPlus pH sensors
Hazardous LocationsClass 1 Div 2, Groups A, B, C, D (in combination with approved sensors)
Keypad TypeTactile keypad with 7-key menu navigation
Max. Relative Humidity95%, non-condensing
Mounting OptionsWall, panel or pipe
Outputs2 x 4-20 mA outputs 3 relays 240 VAC 50 Hz single phase
RangeSensor Dependant
Relay DetailsThree form C relays, 5A @ 250VAC
ResolutionSensor Dependant
Temperature (English) Operating-4°F to +140°F
USB InterfaceFront-panel accessible USB-A for firmware updates and datalog downloads
Voltage90/240 V
Weight (English)3 lbs
Weight (Metric)1.3 kg
Temperature (Metric) Operating-20°C to +60°C
TypeProcess Analyzer
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link