Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Immunogen sequence: RKLTKNSEADGMFAY
Reconstitution: 25 uL, 50 uL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
This gene encodes a transmembrane protein component of a mechanosensitve ion channel that is activated by mechanical stimuli in various cell types and confers slowly adapting, mechanically activated currents in dorsal root ganglion neurons. Mechanically activated ion channels are sensors that are critical for hearing, touch, pain, and blood pressure regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tentonin 3; Transmembrane protein 150C
Gene Aliases: RGD1306105; Tmem150c; Ttn3
UniProt ID: (Rat) B5DFH9, (Mouse) Q8C8S3
Entrez Gene ID: (Rat) 360916, (Mouse) 231503
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support