APIX/APIMS δQ and APIX/APIMS Quattro™ Ultra High Purity (UHP) Electronic Gas Analyzers
APIX/APIMS δQ and APIX/APIMS  Quattro™ Ultra High Purity (UHP) Electronic Gas Analyzers
Thermo Scientific™
APIX/APIMS δQ and APIX/APIMS Quattro™ Ultra High Purity (UHP) Electronic Gas Analyzers
Perform routine continuous measurement of contaminants including water and oxygen to single-digit ppt levels with these gas analyzers.
Have Questions?
Catalog NumberProduct Type
APIMSXANLYZUltra High Purity Gas Analyzer
Catalog number APIMSXANLYZ
Price (USD)
-
Request A Quote
Product Type:
Ultra High Purity Gas Analyzer
Ask our AI about this Product
Perform routine continuous measurement of contaminants including water and oxygen to single-digit ppt levels with Thermo Scientific™ APIX/APIMS δQ and APIX/APIMS Quattro Ultra High Purity (UHP) Electronic Gas Analyzers.
These gas analyzers offer a cost effective alternative to conventional quality control techniques allowing each bulk gas to be monitored for a range of potential contaminants, while achieving lower detection limits (up to 100 times better) than any technology.
  • Reliable, fully automatic system for online quality control
  • Superior sensitivity with lower detection capability of <10 ppt
  • Automatic switching and calibration
  • Fast online measurement (typically <5 seconds for complete analysis)
  • Industry standard communications protocol for plant control and integration
Recommended for:
  • UHP Nitrogen
  • UHP Argon
  • UHP Helium
  • UHP Hydrogen
Specifications
Analysis TimeCycle time 2 minutes
Bulk Gas SuitabilityH2, N2, Ar, He (plus O2 for dB model)
DescriptionElectronic gas analyzer
Detection Noise100 MHz (pulse counting type)
Hertz100 MHz (pulse counting type)
Impurities MonitoredH2, CH4, H2O, CO, N2, O2, Ar, CO2, Kr and Xe (depending on bulk gas)
Ion Source Background<1ppt
Measurement MethodAPIMS dB: 1x Triple filter; APIMS Quattro: 4x Triple filter
Serial ConnectionsRS232 Serial Port
Detection Limits<10 ppt (component dependent)
Detector TypePulse Counting Channel Electron Multiplier
Ion Source TypeAtmospheric Pressure Ionization
Product TypeUltra High Purity Gas Analyzer
Unit SizeEach
access the user manuals for mass spectrometry instruments
To access the user manuals of your online mass spectrometer, please contact us directly:
email: Techsupport.epm.europe@thermofisher.com
phone:  +44 1606 548 753
Service Offerings
  • Field Service Support
  • Technical Support
  • Repairs & Calibrations
  • Technical Training
  • Spare Parts Catalogs
  • Promotions & much more…
Purchase Spare Parts
Order Spare parts 24/7 via credit card or PO
(United States customers only)
Factory Service Depot
Request an RMA number for a repair or calibration at one of our facilities
Signup NOW for our quarterly service newsletter for the latest news & updates!
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Frequently asked questions (FAQs)
Information is provided to aid in the running of Gasworks Software in a 21 CFR Part 11 compliant environment. It should be noted that system qualification, validation, and implementation of standard operating procedures are essential elements of Part 11 compliance that cannot be achieved through software functionality alone. While many of the requirements in the regulations are addressed by Thermo Fisher Scientific software features, all aspects of the system must be validated by the customer or their designee.
The Modbus Register Mapping Wizard requires the operator to respond to a series of questions regarding the data to be communicated and the address area to be reserved. This becomes an editable table of values.
A number of different user groups are defined by the system administrator. Each group can be provided with a configurable set of access permissions. The administrator is given the option of limiting the number of password retries, assigning an expiration date for passwords, enabling an audit trail, and setting automatic log out.
The System Wizard fully documents the analyzer hardware. This electronic manual allows the operator to drill down to a component of interest. A brief explanation of selected item is provided with tutorial views and a parts manual.
GasWorks Software has its own data review program that can be installed on an office PC and reviewed offline.
Share catalog number, name or link
1x1 image pixel for data collection