Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Thermo Scientific™

Orion™ AquaMate Vis and UV-Vis Spectrophotometers

Optimized for your water analysis needs AquaMate 7100 Vis and AquaMate 8100 UV-Vis Spectrophotometers provide fast and reliable results
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypePlug TypeSpectral Bandwidth
AQ81008100 UV-Vis SpectrophotometerNA, EU, UK Power cord2 nm
AQ71007100 Vis SpectrophotometerNA, EU, UK Power cord5 nm
AQ7100APAC7100 Vis SpectrophotometerChina, India, ANZ Power Cord5 nm
AQ8100APAC8100 UV-Vis SpectrophotometerChina, India, ANZ Power Cord2 nm
Catalog number AQ8100
Price (USD)
9,090.00
Online Exclusive
10,100.00
Save 1,010.00 (10%)
Each
Estimated availability date  Pending  
Add to cart
Type:
8100 UV-Vis Spectrophotometer
Plug Type:
NA, EU, UK Power cord
Spectral Bandwidth:
2 nm
Recurring order eligible. Learn more »
Price (USD)
9,090.00
Online Exclusive
10,100.00
Save 1,010.00 (10%)
Each
Add to cart
Thermo Scientific™ Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories. AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation.

Pre-programmed Methods:

  • More than 260 preprogrammed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries

Versatility:

  • Glove friendly 7-inch touch screen controller
  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm OD

Verification:

  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further
Specifications
Baseline Flatness±0.002 A
Certifications/ComplianceCE / TUV
CompartmentsAccessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height Removable, washable sample compartment liner with magnetic placement and hold-down
ConnectionsSingle USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Depth (Metric)35.5 cm
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift<0.0005 A/hr. (At 500 nm after 1 hour warmup)
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
For Use With (Application)Water and Waste Water Analysis
Height (Metric)19.5 cm
IncludesTest Tube Holder for vials and test tube from 12 to 25 mm OD, 10 mm square single-cell holder, AquaMate Water Analysis Methods Software, Snap-on Printer, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362).
InterfaceGlove friendly touch-screen
KeypadGlove- friendly touchscreen
Lamp Life>5 years typical
Length (Metric)35.5 cm
Noise(RMS at 500 nm 60 consecutive measurements) ≤0.00020A at 0A at 260 and 500 nm ≤0.00030A at 1A at 260 and 500 nm ≤0.00040A at 2A at 260 and 500 nm
Optical DesignDouble Beam
Photometric Accuracy±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A
Photometric Display-3A to +5A
Photometric Range-2A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Plug TypeNA, EU, UK Power cord
Printer IncludedSnap-on printer included
Scan SpeedSlow, medium and fast - up to 1600 nm/min.
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Warranty1 Year
Wavelength Accuracy±0.5 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Weight (Metric)7.5 kg
Width (Metric)38.5 cm
Detector TypeDual Silicon Photodiodes
Display7 in. High Definition Color Touchscreen, Fixed, 800 x 1280 pixels
Spectral Bandwidth2 nm
Type8100 UV-Vis Spectrophotometer
Wavelength Range190 to 1100 nm
Unit SizeEach
software-resource
Generating response

Videos

Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
 
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Share catalog number, name or link

      1x1 image pixel for data collection