Thermo Scientific™

AquaTrace™ ppb Dissolved Oxygen System

Catalog number: AQD1A0
Have Questions?
Thermo Scientific™

AquaTrace™ ppb Dissolved Oxygen System

Catalog number: AQD1A0
Have Questions?

Monitor trace oxygen levels in critical processes to allow quick response and minimize costly downtime with the Thermo Scientific™ AquaTrace™ ppb Dissolved Oxygen System, a trace dissolved oxygen sensor that measures accurately in pure water processes. Best performance is achieved when used in applications where process temperature, flow and pressure are stable.

 
Catalog Number
AQD1A0
Unit Size
Each
Voltage
24 VDC
Price (USD)
Full specifications
Communication DetailsModbus R TU, DeviceNet, CANopen, EtherNet, USB, RJ-45, RS485
DescriptionIntegral AV38 with one current output, flow chamber
Temperature (Metric) Operating5°C to 50°C
TypeDissolved Oxygen System
Mounting Options1 in. NPT threads at back of sensor, 1 in. NPT threads at front of sensor
Outputs1
Regulatory DetailsRoHS compliant assembly
Temperature (English) Operating41°F to 122°F
Voltage24 VDC
Unit SizeEach
Catalog Number
Unit Size
Each
Voltage
100 to 240 VDC
Price (USD)
Full specifications
Communication DetailsModbus R TU, DeviceNet, CANopen, EtherNet, USB, RJ-45, RS485
Descriptionntegral AV38 with two current outputs, two relays, flow chamber
Temperature (Metric) Operating5°C to 50°C
TypeDissolved Oxygen System
Mounting Options1 in. NPT threads at back of sensor, 1 in. NPT threads at front of sensor
Outputs1
Regulatory DetailsRoHS compliant assembly
Relay Details2
Temperature (English) Operating41°F to 122°F
Voltage100 to 240 VDC
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeVoltagePrice (USD)
AQD1A0Full specifications
Each24 VDCRequest A Quote
Communication DetailsModbus R TU, DeviceNet, CANopen, EtherNet, USB, RJ-45, RS485
DescriptionIntegral AV38 with one current output, flow chamber
Temperature (Metric) Operating5°C to 50°C
TypeDissolved Oxygen System
Mounting Options1 in. NPT threads at back of sensor, 1 in. NPT threads at front of sensor
Outputs1
Regulatory DetailsRoHS compliant assembly
Temperature (English) Operating41°F to 122°F
Voltage24 VDC
Unit SizeEach
AQD1C0Full specifications
Each100 to 240 VDCRequest A Quote
Communication DetailsModbus R TU, DeviceNet, CANopen, EtherNet, USB, RJ-45, RS485
Descriptionntegral AV38 with two current outputs, two relays, flow chamber
Temperature (Metric) Operating5°C to 50°C
TypeDissolved Oxygen System
Mounting Options1 in. NPT threads at back of sensor, 1 in. NPT threads at front of sensor
Outputs1
Regulatory DetailsRoHS compliant assembly
Relay Details2
Temperature (English) Operating41°F to 122°F
Voltage100 to 240 VDC
Unit SizeEach
Showing 2 of 2

Features:

  • Reagent-free amperometric design
  • Trace DO system includes Trace DO Thermo Scientific™ DataStick™, flow cell, AV38 local display; assembled on the panel for simple installation
  • Temperature compensated
  • Pre-calibrated, plug & play sensors
  • Rugged membrane in replaceable sensor cap
  • Remote measurement, calibration, configuration and diagnostics
  • Convenient turn-key AquaTrace DO monitoring system for reliable low DO measurements with a small footprint

Markets/Applications:

  • Process water, production and distribution: Collect quality control data
  • Food and beverage: Monitor process water quality
  • Pure water analysis: Accurate trace DO measurements and minimal drift

DataStick:

  • Provides universal conversion of sensor signals and interactive communications for measurement, calibration, configuration and diagnostics

Communications Adapter:

  • Plugs into the DataStick to provide power and direct interactive communications with control systems

AV38 Local Display:

  • Two-line display and seven-key navigation
  • Data reporting with up to two current outputs
  • Two Form C relays
  • Digital communications
  • AquaTrace ppb DO System with Trace DO ppb DataStick and flow cell installed
  • Trace ppb Dissolved Oxygen Sensor Head Pre-calibrated for ppb level dissolved oxygen and temperature
  • Can be plugged into any DataStick to yield accurate 24-bit data

AquaTrace ppb DO System:

  • Trace DO ppb DataStick and flow cell installed
Trace ppb Dissolved Oxygen Sensor Head:
  • Precalibrated for ppb level dissolved oxygen and temperature
  • Can be plugged into any DataStick to yield accurate 24-bit data

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text