Applied Biosystems™

Pathatrix™ Sample Vessel Holder

Catalog number: ATUBEHOLD
Have Questions?
Applied Biosystems™

Pathatrix™ Sample Vessel Holder

Catalog number: ATUBEHOLD
Have Questions?
Catalog Number
ATUBEHOLD
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ATUBEHOLD1 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Pathatrix™ Sample Vessel Holder is designed for use with the Pathatrix™ Auto Instrument. The apparatus holds the Sample Vessel and Elution Vessel and facilitates easy loading of the vessels with the sample and elution solution, respectively. The Elution Vessel holder features a magnet positioned for the rapid capture of the paramagnetic particles.

Specifications

Capacity
1 Sample Vessel and Elution Vessel Assembly
For Use With (Equipment)
Pathatrix™ Auto System
Product Line
Pathatrix™
Shipping Condition
Room Temperature
Quantity
1 each
Product Type
Sample Vessel Holder
Unit Size
1 each

Contents & Storage

1 holder

Store at Room Temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text