Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Enclosure Rating | IP65 (NEMA 4X) |
Input Power | 10 VDC to 30 VDC |
Live Density Input | UGC, 4 to 20 mA |
Program Memory | 256 MB |
Audit Trails | User configurable; defaulting to 200 audit events, 60 different types of audits |
Certifications/Compliance | CSA/C-US Class I, Div 2, Groups C and D hazardous locations; ambient temperature range of -40°C to +85°C, temperature code T3Cl Type 4X enclosure) |
Communication Ports | (2) Serial (2) Ethernet (1) USB |
Depth (English) | 6.9 in. |
Depth (Metric) Cover | 175.26 mm |
Height (English) | 7.5 in. |
Height (Metric) | 190.5 mm |
Operating Humidity Range | 0 to 95% RH, Non-condensing |
Width (English) | 7.18 in. |
Width (Metric) | 182.37 mm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
AUTOFLEX also known as AUTOFLEXFLOWCOMPUTER | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|