Thermo Scientific™
Sensititre™ Avian AVIAN1F Vet AST Plate
Catalog number: AVIAN1F
Have Questions?
Thermo Scientific™
Sensititre™ Avian AVIAN1F Vet AST Plate
Catalog number: AVIAN1F
Have Questions?
Perform accurate AST with an avian- and poultry-specific formulary using Thermo Scientific™ Sensititre™ Avian AVIAN1F AST Plate. Sensititre Avian AVIAN1F AST Plate delivers accurate results on a single plate format reducing the need for offline tests and retests.
Catalog Number
AVIAN1F
Unit Size
Each
Specimen Type
One-Isolate, Avian
Target Organism Class
Non-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Quantity
10/Pk.
Price (USD)
Full specifications
Antimicrobial AgentsVarious
Antibiotic CodeVarious
Product TypeMicrobiology Media
Target Organism ClassNon-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test TypeVeterinary AST
Isolates Per Plate1
Quantity10/Pk.
Reading OptionsAutoread or manual
Specimen TypeOne-Isolate, Avian
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeSpecimen TypeTarget Organism ClassQuantityPrice (USD)
AVIAN1FFull specifications
EachOne-Isolate, AvianNon-fastidious, Gram-positive and Gram-negative Veterinary Isolates10/Pk.Request A Quote
Antimicrobial AgentsVarious
Antibiotic CodeVarious
Product TypeMicrobiology Media
Target Organism ClassNon-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test TypeVeterinary AST
Isolates Per Plate1
Quantity10/Pk.
Reading OptionsAutoread or manual
Specimen TypeOne-Isolate, Avian
Unit SizeEach
Showing 1 of 1
Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Avian AVIAN1F AST Plate.
  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Automated or manual read options are available for ultimate flexibility.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For Veterinary Use Only.