20X Bolt™ MOPS SDS Running Buffer
20X Bolt™ MOPS SDS Running Buffer
Invitrogen™

20X Bolt™ MOPS SDS Running Buffer

Bolt MOPS SDS Running Buffer (20X) is formulated for running proteins on Bolt Bis-Tris Plus gels. It is recommended forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
B0001500 mL
B0001025 L
Catalog number B0001
Price (USD)
111.65
Online Exclusive
117.00
Save 5.35 (5%)
Each
-
Add to cart
Quantity:
500 mL
Price (USD)
111.65
Online Exclusive
117.00
Save 5.35 (5%)
Each
Add to cart
Ask our AI about this Product
Bolt MOPS SDS Running Buffer (20X) is formulated for running proteins on Bolt Bis-Tris Plus gels. It is recommended for separating medium- to large-sized proteins.

Use the right buffer to optimize protein separations
Bolt MES SDS Running Buffer and Bolt MOPS SDS Running Buffer can both be used with Bolt Bis-Tris Plus gels. MES has a lower pKa than MOPS, making the MES SDS Running Buffer faster than the MOPS SDS running buffers. The difference in ion migration affects stacking and results in a difference in protein separation range between these buffers. Use of MOPS buffer allows proteins to run slower than when using MES buffer.

Compare protein migration patterns using MES and MOPs on Bolt Bis-Tris Plus gels

See all available buffers and reagents available for SDS-PAGE

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialRunning Buffer
Recommended StorageMOPS SDS Running Buffer (20X).

Store at room temperature.

Shelf-life is 6 months, when stored properly.
Concentration20X
For Use With (Equipment)Bolt
Physical FormLiquid
Product LineBolt
Quantity500 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link