20X Bolt™ MES SDS Running Buffer
20X Bolt™ MES SDS Running Buffer
Invitrogen™

20X Bolt™ MES SDS Running Buffer

Bolt MES SDS Running Buffer (20X) is formulated for running proteins on Bolt Bis-Tris gels. It is recommended for separatingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
B0002025 L
B0002500 mL
Catalog number B000202
Price (USD)
680.65
Online Exclusive
704.00
Save 23.35 (3%)
Each
In stock
Add to cart
Quantity:
5 L
Recurring order eligible. Learn more »
Price (USD)
680.65
Online Exclusive
704.00
Save 23.35 (3%)
Each
Add to cart
Ask our AI about this Product
Bolt MES SDS Running Buffer (20X) is formulated for running proteins on Bolt Bis-Tris gels. It is recommended for separating small- to medium-sized proteins.

Use the right buffer to optimize protein separations
Bolt MES SDS Running Buffer and Bolt MOPS SDS Running Buffer can both be used with Bolt Bis-Tris gels. MES has a lower pKa than MOPS, making the MES SDS Running Buffer faster than the MOPS SDS running buffers. The difference in ion migration affects stacking and results in a difference in protein separation range between these buffers. Use of MOPS buffer allows proteins to run slower than when using MES buffer.

Compare protein migration patterns using MES and MOPs on Bolt Bis-Tris Plus gels

See all available buffers and reagents available for SDS-PAGE

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialRunning Buffer
FormulationSodiumdodecyl Sulphate 151-21-3 ( 1-5 )
Recommended StorageMES SDS Running Buffer (20X).

Store at 4°C to 25°C.

Shelf-life is 6 months, when stored properly.
Concentration20X
For Use With (Application)Protein electrophoresis & western blotting
For Use With (Equipment)Bolt
Physical FormLiquid
Product LineBolt
Quantity5 L
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Are Bolt gels compatible with other transfer devices?
Are Bolt gels compatible with transfer devices from other suppliers?
Is there a difference in composition between the NuPAGE MES SDS Running Buffer (20X) and the 20X Bolt MES SDS Running Buffer?
Which power supplies are compatible with the Mini Gel Tank?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

What are the storage conditions for Bolt gels?

Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the shelf life for Bolt gels?

The shelf life for Bolt gels is similar to that for NuPAGE gels: 12 months from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

Videos

How To Run the Bolt Pre-Cast Protein Gel
How to run the Bolt Bis-Tris Plus Pre-cast Protein gel on the Bolt Mini-Gel Tank. Do you run pre-cast protein gels? Western blotting? If you do, you should check out the New Bolt Pre-Cast Protein Gels and Bolt Electrophoresis Tank.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
 
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Customers who viewed this item also viewed



      Documents & Downloads

      Certificates

      Lot #Certificate TypeDateCatalog Number(s)
      3233149Certificate of AnalysisJul 19, 2025B000202
      3223763Certificate of AnalysisJun 27, 2025B0002
      3218903Certificate of AnalysisJun 24, 2025B000202
      3167871Certificate of AnalysisJun 10, 2025B000202
      3178555Certificate of AnalysisMay 22, 2025B0002
      5 results displayed, search above for a specific certificate

      Safety Data Sheets

      Share catalog number, name or link