Invitrogen™

Biotin Azide (PEG4 carboxamide-6-Azidohexanyl Biotin)

Catalog number: B10184
Have Questions?
Invitrogen™

Biotin Azide (PEG4 carboxamide-6-Azidohexanyl Biotin)

Catalog number: B10184
Have Questions?
Catalog Number
B10184
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B101841 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The hapten, biotin azide is reactive with terminal alkynes via a copper-catalyzed click reaction. Biotin can be subsequently detected with streptavidin, avidin or NeutrAvidin® biotin-binding protein.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Solid
Quantity
1 mg
Product Type
Biotin Azide
Molecular Weight (g/mol)
615.79 Da
Solubility
DMSO (Dimethylsulfoxide)
Chemical Reactivity
Alkyne
Reactive Group
Azide
Shipping Condition
Room Temperature
Reactive Moiety
Amine, Azide
Label or Dye
Biotin
Label Type
Biotin and Other Haptens
Color
None
Product Line
Molecular Probes™

Contents & Storage

Store at ≤-20°C and keep desiccated.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text