Invitrogen™

BODIPY™ FL Maleimide (BODIPY™ FL N-(2-Aminoethyl))Maleimide)

Catalog number: B10250
Have Questions?
Invitrogen™

BODIPY™ FL Maleimide (BODIPY™ FL N-(2-Aminoethyl))Maleimide)

Catalog number: B10250
Have Questions?
Catalog Number
B10250
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B102505 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The thiol-reactive BODIPY™ FL maleimide produces electronically neutral dye conjugates that are spectrally similar to the negatively charged fluorescein dye. This dye's lack of ionic charge results in minimal effects on the isoelectric points of standard proteins conjugated with this fluorophore. The small size and relatively long excited-state lifetime of BODIPY™ FL dye has proven useful for studying ligand-receptor interaction by fluorescence polarization. In addition, BODIPY™ FL dye has little or no spectral overlap with longer-wavelength dyes such as tetramethylrhodamine and Texas Red™ dye, making it a useful green fluorophore for multicolor applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
5 mg
Product Type
Maleimide
Chemical Reactivity
Thiol
Shipping Condition
Room Temperature
Reactive Moiety
Maleimide
Label or Dye
BODIPY™ FL
Label Type
BODIPY Dyes
Product Line
BODIPY™

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text