Invitrogen™

BAPTA, Tetracesium Salt, cell impermeant

Catalog number: B1212
Have Questions?
Invitrogen™

BAPTA, Tetracesium Salt, cell impermeant

Catalog number: B1212
Have Questions?
Catalog Number
B1212
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B12121 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
BAPTA is a cell-impermeant chelator, which is a highly selective for Ca2+ over Mg2+, and it can be used to control the level of extracellular Ca2+. BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH.

Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Cell Viability & Proliferation
Dye Type
Chelator (Non-Fluorescent)
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text