Bovine Serum Albumin (BSA)
Bovine Serum Albumin (BSA)
Thermo Scientific™

Bovine Serum Albumin (BSA)

Thermo Scientific Bovine Serum Albumin (BSA) is ideal for stabilization of enzymes during storage and for enzymatic reactions where theRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
B14Promo Image5 mg
Catalog number B14
Price (USD)
18.65
Online Exclusive
22.84
Save 4.19 (18%)
Each
In stock
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)
18.65
Online Exclusive
22.84
Save 4.19 (18%)
Each
Add to cart
Thermo Scientific Bovine Serum Albumin (BSA) is ideal for stabilization of enzymes during storage and for enzymatic reactions where the absence of nucleases is essential. BSA increases PCR yields from low purity templates. It also prevents adhesion of enzymes to the reaction tubes and tip surfaces.

Highlights

• Non-modified (non-acetylated)
• Fraction V

Applications

• Enzyme stabilization
• PCR with low purity templates, e.g. mouse tail DNA
• Restriction enzyme digestion
For Research Use Only. Not for use in diagnostic procedures.
Specifications
GradeMolecular Biology
Product TypeBovine Serum Albumin
Quantity5 mg
Concentration20 mg/mL
Research CategoryTraditional Cloning
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link