Invitrogen™

Biocytin Hydrazide

Catalog number: B1603
Have Questions?
Invitrogen™

Biocytin Hydrazide

Catalog number: B1603
Have Questions?
Catalog Number
B1603
also known as B-1603
Unit Size
25 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B1603
also known as B-1603
25 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Biocytin hydrazide an be used as microinjectable anterograde and transneuronal tracers that is fixable with aldehyde-based fixatives. Additionally, the primary aliphatic amine of this molecule can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Biocytin Hydrazide
Quantity
25 mg
Reagent Type
Biocytins
Shipping Condition
Room Temperature
Label Type
Biotin & Other Haptens

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text