Invitrogen™

X-Gal (5-Bromo-4-Chloro-3-Indolyl β-D-Galactopyranoside)

Catalog number: B1690
Have Questions?
Invitrogen™

X-Gal (5-Bromo-4-Chloro-3-Indolyl β-D-Galactopyranoside)

Catalog number: B1690
Have Questions?
Catalog Number
B1690
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B16901 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
X-Gal is a widely used chromogenic substrate for β-galactosidase. It yields a dark blue precipitate at the site of enzymatic activity. X-Gal is useful for numerous histochemical and molecular biology applications, including detection of lacZ activity in cells and tissues. In contrast to β-glucuronidase as a gene marker, β-galactosidase can be fixed in cells and tissues with glutaraldehyde without loss of activity and detected with high resolution using X-Gal.


• Chromogenic substrate
• Cell permeant
• β-galactosidase activity can still be detected using X-Gal after fixation


Additional Information About Glycosidase Enzymes
Glycosidase enzymes exhibit very high selectivity for hydrolysis of their preferred sugars. For example, β-galactosidase rapidly hydrolyzes β-D-galactopyranosides but usually does not hydrolyze either the anomeric α-D-galactopyranosides or the isomeric β-D-glucopyranosides. Endogenous glycosidase activity is frequently used to characterize strains of microorganisms and to selectively label organelles of mammalian cells; defects in glycosidase activity are characteristic of several diseases.

In addition, glycosidases are important reporter gene markers. Specifically, lacZ, which encodes β-galactosidase, is extensively used as a reporter gene in animals and yeast, whereas the β-glucuronidase (GUS) gene is a popular reporter gene in plants. Glycosidase substrates are also used in conjunction with glycosidase-conjugated secondary detection reagents in immunohistochemical techniques and enzyme-linked immunosorbent assays (ELISAs). See all of our Molecular Probes™ fluorogenic and chromogenic glycosidase substrates.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Permeability
Cell-Permeant
Detection Method
Colorimetric
For Use With (Application)
Molecular Biology
Quantity
1 g
Substrate
Beta-Gal Substrate
Substrate Properties
Chemical Substrate
Substrate Type
Beta-Gal Substrate
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text