Thermo Scientific Chemicals

4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B20015.18
50 g, Each
Thermo Scientific Chemicals

4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B20015.18
50 g, Each
Quantity
Catalog number: B20015.18
also known as B20015-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17852-52-7
IUPAC Name
hydrogen 4-hydrazinylbenzene-1-sulfonamide chloride
Molecular Formula
C6H10ClN3O2S
InChI Key
IKEURONJLPUALY-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=C(C=C1)S(N)(=O)=O
Appearance (Color)
Pale cream to cream to pale brown
Form
Crystals or powder or crystalline powder or flakes
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text