Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 97%, Thermo Scientific Chemicals
Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B20028.06
also known as B20028-06
Price (USD)/ Each
395.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
395.00
Add to cart
Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberB20028.06
Price (USD)/ Each
395.00
-
Add to cart
Chemical Identifiers
CAS121071-71-4
IUPAC Name2,3-dimethyl 4-aminothiophene-2,3-dicarboxylate
Molecular FormulaC8H9NO4S
InChI KeyXQPDCUCAKYNVAO-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C(=O)OC)C(N)=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale orange or pale pink
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only